Imprimer

Anti-IgG Goat Polyclonal Antibody (CF™ 660C)

Fournisseur: Biotium
Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5


20052 20052-1
20052.EA 347 EUR
20052. 20052-1
Anti-IgG Goat Polyclonal Antibody (CF™ 660C)
Anticorps
Type: Secondary
Antigen: IgG (Mouse) (mouse IgG)
Clonality: polyclonal
Clone:
Conjugation: CF® 660C Dye
Epitope:
Host: Goat
Isotype: none
Reactivity: Mouse
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR