Imprimer

Anti-IgG Rabbit Polyclonal Antibody (CF™ 568)

Fournisseur: Biotium
Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5


20107 20107-1
20107.EA 289 EUR
20107. 20107-1
Anti-IgG Rabbit Polyclonal Antibody (CF™ 568)
Anticorps
Type: Secondary
Antigen: IgG (Goat) (goat IgG)
Clonality: polyclonal
Clone:
Conjugation: CF® 568 Dye
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Goat
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR