Imprimer

Anti-IgG Goat Polyclonal Antibody (CF™ 594)

Fournisseur: Biotium
Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

20110.EA 304 EUR
20110. 20110-1
Anti-IgG Goat Polyclonal Antibody (CF™ 594)
Anticorps
Type: Secondary
Antigen: IgG (Mouse) (mouse IgG)
Clonality: polyclonal
Clone:
Conjugation: CF® 594 Dye
Epitope:
Host: Goat
Isotype: none
Reactivity: Mouse
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR