Imprimer

Anti-IgG Rabbit Polyclonal Antibody (CF™ 633)

Fournisseur: Biotium
Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

20128-1EA 122 EUR
20128-1 20128.
Anti-IgG Rabbit Polyclonal Antibody (CF™ 633)
Anticorps
Type: Secondary
Antigen: IgG (Goat) (goat IgG)
Clonality: polyclonal
Clone:
Conjugation: CF® 633 Dye
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Goat
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR