Imprimer

Anti-IgG Goat Polyclonal Antibody (CF™ 350)

Fournisseur: Biotium
Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5


20144.EA 347 EUR
20144. 20144-1
Anti-IgG Goat Polyclonal Antibody (CF™ 350)
Anticorps
Type: Secondary
Antigen: IgG (Rabbit) (rabbit IgG)
Clonality: polyclonal
Clone:
Conjugation: CF® 350 Dye
Epitope:
Host: Goat
Isotype: none
Reactivity: Rabbit
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR