Imprimer
Anti-IgG2b Goat Polyclonal Antibody (CF™ 640R)
Fournisseur: Biotium
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
20271 20271-1
20271.EA 372 EUR
20271. 20271-1
Anti-IgG2b Goat Polyclonal Antibody (CF™ 640R)
Anticorps
Type: Secondary
Antigen: IgG2b (Mouse) (mouse IgG2b)
Clonality: polyclonal
Clone:
Conjugation: CF® 640R Dye
Epitope:
Host: Goat
Isotype: none
Reactivity: Mouse