Imprimer

Anti-Dopamine Receptor D1 Rabbit Polyclonal Antibody

Fournisseur: BosterBio
Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

PA1231
BSBTPA1231EA 0 EUR
BSBTPA1231
Anti-Dopamine Receptor D1 Rabbit Polyclonal Antibody
Anticorps
Type: Primary
Antigen: Dopamine Receptor D1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: near the C-terminus
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR