Imprimer

Anti-Caspase-8(P18) Rabbit Polyclonal Antibody

Fournisseur: Boster
Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

BSBTPA1308EA 550 EUR
BSBTPA1308
Anti-Caspase-8(P18) Rabbit Polyclonal Antibody
Anticorps
Type: Primary
Antigen: Caspase-8(P18)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR