Imprimer

Anti-PI 3 Kinase p85 alpha Rabbit Polyclonal Antibody

Fournisseur: Boster
Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

BSBTPA1410EA 550 EUR
BSBTPA1410
Anti-PI 3 Kinase p85 alpha Rabbit Polyclonal Antibody
Anticorps
Type: Primary
Antigen: PI 3 Kinase p85 alpha
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: near the N-terminus
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR