Imprimer
Anti-Podoplanin/gp36 Rabbit Polyclonal Antibody
Fournisseur: Boster
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
BSBTPA1411EA 550 EUR
BSBTPA1411
Anti-Podoplanin/gp36 Rabbit Polyclonal Antibody
Anticorps
Type: Primary
Antigen: Podoplanin/gp36
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: near the C-terminus
Host: Rabbit
Isotype: IgG
Reactivity: Human