Imprimer

Anti-STAT2 Rabbit Polyclonal Antibody

Fournisseur: BosterBio
Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

PA1509
BSBTPA1509EA 550 EUR
BSBTPA1509
Anti-STAT2 Rabbit Polyclonal Antibody
Anticorps
Rabbit IgG polyclonal antibody for Signal transducer and activator of transcription 2(STAT2) detection. Tested with WB in Human.

Reconstitute with distilled water.

Add 0,2 ml of distilled water will yield a concentration of 500 µg/ml.

Type: Primary
Antigen: STAT2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: near the C-terminus
Host: Rabbit
Isotype: IgG
Reactivity: Human
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR