Imprimer

Anti-LIF Rabbit Polyclonal Antibody

Fournisseur: Boster
Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

BSBTPA1562EA 550 EUR
BSBTPA1562
Anti-LIF Rabbit Polyclonal Antibody
Anticorps
Type: Primary
Antigen: LIF
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: near the C-terminus
Host: Rabbit
Isotype: IgG
Reactivity: Mouse, Rat
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR