Imprimer

Anti-NF-kB p65 Rabbit Polyclonal Antibody

Fournisseur: Boster
Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

BSBTPA1669EA 550 EUR
BSBTPA1669
Anti-NF-kB p65 Rabbit Polyclonal Antibody
Anticorps
Rabbit IgG polyclonal antibody for Transcription factor p65(RELA) detection. Tested with WB, IHC-P, ICC in Human;Mouse;Rat.

RELA(V-REL AVIAN RETICULOENDOTHELIOSIS VIRAL ONCOGENE HOMOLOG A), also called NFKB3 or NFKB, p65 SUBUNIT. NFKB1 or NFKB2 is bound to REL, RELA, or RELB to form the NFKB complex. The NFKB complex is inhibited by I-kappa-B proteins, which inactivate NFKB by trapping it in the cytoplasm. The p65(RELA) heterodimer is the most abundant form of NFKB. And the RELA gene is located on 11q13.1. RELA is a nonhistone substrate of HDAC3 and that IKBA-dependent nuclear export of the HDAC3-deacetylated RELA replenishes the depleted cytoplasmic pool of latent NFKB-IKBA complexes for subsequent NFKB responses. RELA nucleocytoplasmic redistribution coincided with export of PPARG, and immunoprecipitation analysis indicated that PPARG-RELA association was dependent on the PPARG C-terminal ligand-binding domain. IKK-dependent phosphorylation of RELA on ser468 enhanced binding of GCN5 to RELA and RELA ubiquitination.

Add 0.2ml of distilled water will yield a concentration of 500ug/ml.

Each vial contains 5mg BSA, 0.9mg NaCl, 0.2mg Na2HPO4, 0.05mg Thimerosal, 0.05mg NaN3.

Tested Species: In-house tested species with positive results.
Predicted Species: Species predicted to be fit for the product based on sequence similarities.
By Heat: Boiling the paraffin sections in 10mM citrate buffer, pH6.0, for 20mins is required for the staining of formalin/paraffin sections.
Other applications have not been tested.
Optimal dilutions should be determined by end users.

Type: Primary
Antigen: NF-kB p65
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR