Imprimer

Anti-SDHB Rabbit Polyclonal Antibody

Fournisseur: BosterBio
Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

PA1718
BSBTPA1718EA 550 EUR
BSBTPA1718
Anti-SDHB Rabbit Polyclonal Antibody
Anticorps
Rabbit IgG polyclonal antibody for Succinate dehydrogenase[ubiquinone] iron-sulfur subunit, mitochondrial (SDHB) detection. Tested with WB, IHC-P in Human; Mouse; Rat.

Reconstitute with distilled water.

Add 0,2 ml of distilled water will yield a concentration of 500 µg/ml.

Type: Primary
Antigen: SDHB
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: near the N-terminus
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR