Imprimer

Anti-TLR8 Rabbit Polyclonal Antibody

Fournisseur: BosterBio
Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

PA1734
BSBTPA1734EA 550 EUR
BSBTPA1734
Anti-TLR8 Rabbit Polyclonal Antibody
Anticorps
Rabbit IgG polyclonal antibody for Toll-like receptor 8 (TLR8) detection. Tested with WB in Rat; Mouse.

Reconstitute with distilled water.

Add 0,2 ml of distilled water will yield a concentration of 500 µg/ml.

Type: Primary
Antigen: TLR8
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: near the C-terminus
Host: Rabbit
Isotype: IgG
Reactivity: Rat, Mouse
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR