Imprimer

Anti-Fatty Acid Binding Protein 5 Rabbit Polyclonal Antibody

Fournisseur: BosterBio
Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

PA1853
BSBTPA1853EA 550 EUR
BSBTPA1853
Anti-Fatty Acid Binding Protein 5 Rabbit Polyclonal Antibody
Anticorps
Rabbit IgG polyclonal antibody for Fatty acid-binding protein, epidermal (FABP5) detection. Tested with WB, IHC-P in Mouse; Rat.

Reconstitute with distilled water.

Add 0,2 ml of distilled water will yield a concentration of 500 µg/ml.

Type: Primary
Antigen: Fatty Acid Binding Protein 5
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: near the N-terminus
Host: Rabbit
Isotype: IgG
Reactivity: Mouse, Rat
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR