Imprimer

Anti-NFkB p100/p52 Rabbit Polyclonal Antibody

Fournisseur: Boster
Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

BSBTPA1946-1EA 0 EUR
BSBTPA1946-1
Anti-NFkB p100/p52 Rabbit Polyclonal Antibody
Anticorps
Rabbit IgG polyclonal antibody for Nuclear factor NF-kappa-B p100 subunit (NFKB2) detection. Tested with WB in Human.

Reconstitute with distilled water.

Add 0,2 ml of distilled water will yield a concentration of 500 µg/ml.

Type: Primary
Antigen: NFkB p100/p52
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: near the N-terminus
Host: Rabbit
Isotype: IgG
Reactivity: Human
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR