Imprimer

Anti-NRF1 Rabbit Polyclonal Antibody

Fournisseur: BosterBio
Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

PA1948
BSBTPA1948EA 550 EUR
BSBTPA1948
Anti-NRF1 Rabbit Polyclonal Antibody
Anticorps
Rabbit IgG polyclonal antibody for Nuclear respiratory factor 1 (NRF1) detection. Tested with WB, IHC-P, ICC in Human; Mouse; Rat.

Add 0,2 ml of distilled water will yield a concentration of 500 µg/ml.

Type: Primary
Antigen: NRF1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR