Imprimer

Anti-CYP2U1 Rabbit Polyclonal Antibody

Fournisseur: BosterBio
Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

PA2022
BSBTPA2022EA 550 EUR
BSBTPA2022
Anti-CYP2U1 Rabbit Polyclonal Antibody
Anticorps
Rabbit IgG polyclonal antibody for Cytochrome P450 2U1 (CYP2U1) detection. Tested with WB, IHC-P, ICC in Human.

Reconstitute with distilled water.

Add 0,2 ml of distilled water will yield a concentration of 500 µg/ml.

Type: Primary
Antigen: CYP2U1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: near the C-terminus
Host: Rabbit
Isotype: IgG
Reactivity: Human
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR