Imprimer
Anti-Flotillin 1 Rabbit Polyclonal Antibody
Fournisseur: Boster
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
BSBTPA2033EA 550 EUR
BSBTPA2033
Anti-Flotillin 1 Rabbit Polyclonal Antibody
Anticorps
Type: Primary
Antigen: Flotillin 1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat