Imprimer

Anti-RIP Rabbit Polyclonal Antibody

Fournisseur: BosterBio
Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

PA2051
BSBTPA2051EA 550 EUR
BSBTPA2051
Anti-RIP Rabbit Polyclonal Antibody
Anticorps
Rabbit IgG polyclonal antibody for Receptor-interacting serine/threonine-protein kinase 1 (RIPK1) detection. Tested with WB in Human.

Reconstitute with distilled water.

Add 0,2 ml of distilled water will yield a concentration of 500 µg/ml.

Type: Primary
Antigen: RIP
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR