Imprimer

Anti-Bag3 Rabbit Polyclonal Antibody

Fournisseur: BosterBio
Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

PA2100
BSBTPA2100EA 550 EUR
BSBTPA2100
Anti-Bag3 Rabbit Polyclonal Antibody
Anticorps
Rabbit IgG polyclonal antibody for BAG family molecular chaperone regulator 3(BAG3) detection. Tested with WB, IHC-P in Human, Mouse, Rat.

Reconstitute with distilled water.

Add 0,2 ml of distilled water will yield a concentration of 500 µg/ml.

Type: Primary
Antigen: Bag3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: near the C-terminus
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR