Imprimer

Anti-NAK Rabbit Polyclonal Antibody

Fournisseur: BosterBio
Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

PA2117
BSBTPA2117EA 550 EUR
BSBTPA2117
Anti-NAK Rabbit Polyclonal Antibody
Anticorps
Rabbit IgG polyclonal antibody for Serine/threonine-protein kinase TBK1(TBK1) detection. Tested with WB, IHC-P in Human;Mouse;Rat.

Serine/threonine-protein kinase TBK1, also called TANK-binding kinase 1 or NF-kappa-B-activating kinase is an enzyme that in humans is encoded by the TBK1 gene. The gene was assigned to human chromosome 12q14.2. Serine/threonine kinase plays an essential role in regulating inflammatory responses to foreign agents. TBK1 and NF-kappa-B signaling are essential in KRAS mutant tumors, and established a general approach for the rational identification of codependent pathways in cancer.

Add 0.2ml of distilled water will yield a concentration of 500ug/ml.

Each vial contains 5mg BSA, 0.9mg NaCl, 0.2mg Na2HPO4, 0.05mg Thimerosal, 0.05mg NaN3.

WB: The detection limit for TBK1 is approximately 3ng/lane under reducing conditions.
Tested Species: In-house tested species with positive results.
Predicted Species: Species predicted to be fit for the product based on sequence similarities.
By Heat: Boiling the paraffin sections in 10mM citrate buffer, pH6.0, for 20mins is required for the staining of formalin/paraffin sections.
Other applications have not been tested.
Optimal dilutions should be determined by end users.

Type: Primary
Antigen: NAK
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: near the C-terminus
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR