Imprimer

Anti-HYAL1 Rabbit Polyclonal Antibody

Fournisseur: BosterBio
Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

PA2181-2
BSBTPA2181-2EA 550 EUR
BSBTPA2181-2
Anti-HYAL1 Rabbit Polyclonal Antibody
Anticorps
Rabbit IgG polyclonal antibody for Hyaluronidase-1(HYAL1) detection. Tested with WB in mouse.

Reconstitute with distilled water.

Add 0,2 ml of distilled water will yield a concentration of 500 µg/ml.

Type: Primary
Antigen: HYAL1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: near the C-terminus
Host: Rabbit
Isotype: IgG
Reactivity: Mouse
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR