Imprimer

Anti-IL10 Rabbit Polyclonal Antibody

Fournisseur: BosterBio
Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

RP1015
BSBTRP1015EA 552 EUR
BSBTRP1015
Anti-IL10 Rabbit Polyclonal Antibody
Anticorps
Rabbit IgG polyclonal antibody for Interleukin-10(IL10) detection. Tested with WB, ELISA in Mouse.

Reconstitute with distilled water.

Add 0,2 ml of distilled water will yield a concentration of 500 µg/ml.

Type: Primary
Antigen: IL10
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Mouse
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR