Imprimer

Anti-NTHL1 Rabbit Polyclonal Antibody (DyLight® 550)

Fournisseur: Novus Biologicals
Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

NB100-108R
NOVUNB100-108REA 649 EUR
NOVUNB100-108R
Anti-NTHL1 Rabbit Polyclonal Antibody (DyLight® 550)
Anticorps
Type: Primary
Antigen: NTHL1 (nth endonuclease III-like 1 (E. coli))
Clonality: polyclonal
Clone:
Conjugation: DyLight® 550
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human, Mouse, Rat
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR