Imprimer

Anti-HIF1A Mouse Monoclonal Antibody [clone: H1alpha67] (Biotin)

Fournisseur: Novus Biologicals
Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

NB100-123B
NOVUNB100-123BEA 754 EUR
NOVUNB100-123B
Anti-HIF1A Mouse Monoclonal Antibody [clone: H1alpha67] (Biotin)
Anticorps
Mouse Monoclonal HIF-1 alpha Antibody (H1alpha67) [Biotin]. Tested Applications: Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin. Tested Reactivity: Human, Mouse, Rat, Bovine, Ferret, Primate, Porcine, Sheep.

Recommended Dilutions: Immunohistochemistry (Parafin): 1:500-1:1000

Type: Primary
Antigen: HIF1A
Clonality: Monoclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR