Imprimer

Anti-HIF1A Mouse Monoclonal Antibody (DyLight® 680) [clone: H1alpha67]

Fournisseur: Novus Biologicals
Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

NOVUNB100-123FREA 705 EUR
NOVUNB100-123FR
Anti-HIF1A Mouse Monoclonal Antibody (DyLight® 680) [clone: H1alpha67]
Anticorps
Type: Primary
Antigen: HIF1A (hypoxia inducible factor 1, alpha subunit (basic helix-loop-helix transcription factor))
Clonality: monoclonal
Clone: H1alpha67
Conjugation: DyLight® 680
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity:
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR