Imprimer

Anti-HIF-1 beta Mouse Monoclonal Antibody (Biotin) [clone: H1beta234]

Fournisseur: Novus Biologicals
Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

NOVUNB100-124BEA 739 EUR
NOVUNB100-124B
Anti-HIF-1 beta Mouse Monoclonal Antibody (Biotin) [clone: H1beta234]
Anticorps
Type: Primary
Antigen: HIF-1 beta
Clonality: monoclonal
Clone: H1beta234
Conjugation: Biotin
Epitope:
Host: Mouse
Isotype: IgG1 Kappa
Reactivity:
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR