Imprimer

Anti-EPAS1 Mouse Monoclonal Antibody (DyLight® 550) [clone: ep190b]

Fournisseur: Novus Biologicals
Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

NB100-132R
NOVUNB100-132REA 790 EUR
NOVUNB100-132R
Anti-EPAS1 Mouse Monoclonal Antibody (DyLight® 550) [clone: ep190b]
Anticorps
Type: Primary
Antigen: EPAS1 (endothelial PAS domain protein 1)
Clonality: monoclonal
Clone: ep190b
Conjugation: DyLight® 550
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity:
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR