Imprimer

Anti-CYP19A1 Rabbit Polyclonal Antibody

Fournisseur: Novus Biologicals
Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

NOVUNB100-1596EA 575 EUR
NOVUNB100-1596 NOVUNB100-1596SS
Anti-CYP19A1 Rabbit Polyclonal Antibody
Anticorps
Type: Primary
Antigen: CYP19A1 (cytochrome P450, family 19, subfamily A, polypeptide 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity:
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR