Imprimer

Anti-PECAM1 Rat Monoclonal Antibody (PE (Phycoerythrin)) [clone: MEC 7.46]

Fournisseur: Novus Biologicals
Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

NOVUNB100-1642PEEA 636 EUR
NOVUNB100-1642PE
Anti-PECAM1 Rat Monoclonal Antibody (PE (Phycoerythrin)) [clone: MEC 7.46]
Anticorps
Type: Primary
Antigen: PECAM1 (platelet/endothelial cell adhesion molecule 1)
Clonality: monoclonal
Clone: MEC 7.46
Conjugation: PE (Phycoerythrin)
Epitope:
Host: Rat
Isotype: IgG1
Reactivity: Mouse
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR