Imprimer

Anti-DSG3 Mouse Monoclonal Antibody (DyLight® 550) [clone: 5G11]

Fournisseur: Novus Biologicals
Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

NOVUNB100-1643REA 667 EUR
NOVUNB100-1643R
Anti-DSG3 Mouse Monoclonal Antibody (DyLight® 550) [clone: 5G11]
Anticorps
Type: Primary
Antigen: DSG3 (desmoglein 3)
Clonality: monoclonal
Clone: 5G11
Conjugation: DyLight® 550
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR