Imprimer

Anti-HDAC3 Rabbit Polyclonal Antibody

Fournisseur: Novus Biologicals
Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

NOVUNB100-1669EA 602 EUR
NOVUNB100-1669
Anti-HDAC3 Rabbit Polyclonal Antibody
Anticorps
Type: Primary
Antigen: HDAC3 (histone deacetylase 3)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR