Imprimer
Anti-RAD51C Mouse Monoclonal Antibody (DyLight® 680) [clone: 2H11/6]
Fournisseur: Novus Biologicals
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
NOVUNB100-177FREA 596 EUR
NOVUNB100-177FR
Anti-RAD51C Mouse Monoclonal Antibody (DyLight® 680) [clone: 2H11/6]
Anticorps
Type: Primary
Antigen: RAD51C (RAD51 paralog C)
Clonality: monoclonal
Clone: 2H11/6
Conjugation: DyLight® 680
Epitope:
Host: Mouse
Isotype: IgG1 Kappa
Reactivity:
Commander maintenant