Imprimer

Anti-NKX3-1 Mouse Monoclonal Antibody (DyLight® 488) [clone: 0361]

Fournisseur: Novus Biologicals
Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

NOVUNB100-1828GEA 739 EUR
NOVUNB100-1828G
Anti-NKX3-1 Mouse Monoclonal Antibody (DyLight® 488) [clone: 0361]
Anticorps
Type: Primary
Antigen: NKX3-1 (NK3 homeobox 1)
Clonality: monoclonal
Clone: 361
Conjugation: DyLight® 488
Epitope:
Host: Mouse
Isotype: none
Reactivity: Mouse, Rat
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR