Imprimer

Anti-FANCD2 Rabbit Polyclonal Antibody (DyLight® 405)

Fournisseur: Novus Biologicals
Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

NB100-182V
NOVUNB100-182VEA 782 EUR
NOVUNB100-182V
Anti-FANCD2 Rabbit Polyclonal Antibody (DyLight® 405)
Anticorps
Type: Primary
Antigen: FANCD2 (Fanconi anemia, complementation group D2)
Clonality: polyclonal
Clone:
Conjugation: DyLight® 405
Epitope:
Host: Rabbit
Isotype: none
Reactivity:
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR