Imprimer

Anti-CASR Mouse Monoclonal Antibody [clone: HL 1499]

Fournisseur: Novus Biologicals
Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

NOVUNB100-1830EA 542 EUR
NOVUNB100-1830 NOVUNB100-1830SS
Anti-CASR Mouse Monoclonal Antibody [clone: HL 1499]
Anticorps
Type: Primary
Antigen: CASR (calcium-sensing receptor)
Clonality: monoclonal
Clone: HL 1499
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 Kappa
Reactivity:
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR