Imprimer

Anti-SLC1A3 Rabbit Polyclonal Antibody (FITC (Fluorescein))

Fournisseur: Novus Biologicals
Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

NOVUNB100-1869FEA 606 EUR
NOVUNB100-1869F
Anti-SLC1A3 Rabbit Polyclonal Antibody (FITC (Fluorescein))
Anticorps
Type: Primary
Antigen: SLC1A3 (solute carrier family 1 (glial high affinity glutamate transporter), member 3)
Clonality: polyclonal
Clone:
Conjugation: FITC (Fluorescein)
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Mouse, Rat
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR