Imprimer

Anti-SMN Mouse Monoclonal Antibody [clone: 2B1]

Fournisseur: Novus Biologicals
Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

NOVUNB100-1936SSEA 199 EUR
NOVUNB100-1936SS NOVUNB100-1936
Anti-SMN Mouse Monoclonal Antibody [clone: 2B1]
Anticorps
Type: Primary
Antigen: SMN
Clonality: monoclonal
Clone: 2B1
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity:
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR