Imprimer

Anti-LRP8 Rabbit Polyclonal Antibody (Biotin)

Fournisseur: Novus Biologicals
Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

NOVUNB100-2217BEA 663 EUR
NOVUNB100-2217B
Anti-LRP8 Rabbit Polyclonal Antibody (Biotin)
Anticorps
Type: Primary
Antigen: LRP8 (low density lipoprotein receptor-related protein 8, apolipoprotein e receptor)
Clonality: polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype: none
Reactivity:
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR