Imprimer

Anti-MAP1LC3A Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase))

Fournisseur: Novus Biologicals
Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

NOVUNB100-2220HEA 784 EUR
NOVUNB100-2220H
Anti-MAP1LC3A Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase))
Anticorps
Type: Primary
Antigen: MAP1LC3A (microtubule-associated protein 1 light chain 3 alpha)
Clonality: polyclonal
Clone:
Conjugation: HRP (Horseradish Peroxidase)
Epitope:
Host: Rabbit
Isotype: none
Reactivity:
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR