Imprimer

Anti-S-arrestin Mouse Monoclonal Antibody (HRP (Horseradish Peroxidase)) [clone: PDS-1]

Fournisseur: Novus Biologicals
Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

NOVUNB100-2385HEA 663 EUR
NOVUNB100-2385H
Anti-S-arrestin Mouse Monoclonal Antibody (HRP (Horseradish Peroxidase)) [clone: PDS-1]
Anticorps
Type: Primary
Antigen: S-arrestin
Clonality: monoclonal
Clone: PDS-1
Conjugation: HRP (Horseradish Peroxidase)
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity:
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR