Imprimer

Anti-AGPAT6 Rabbit Polyclonal Antibody (Biotin)

Fournisseur: Novus Biologicals
Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

NB100-2390B
NOVUNB100-2390BEA 538 EUR
NOVUNB100-2390B
Anti-AGPAT6 Rabbit Polyclonal Antibody (Biotin)
Anticorps
Type: Primary
Antigen: AGPAT6 (1-acylglycerol-3-phosphate O-acyltransferase 6)
Clonality: polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype: none
Reactivity:
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR