Imprimer

Anti-RNF5 Mouse Monoclonal Antibody (DyLight® 755) [clone: LHM 2]

Fournisseur: Novus Biologicals
Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

NOVUNB100-2688IREA 596 EUR
NOVUNB100-2688IR
Anti-RNF5 Mouse Monoclonal Antibody (DyLight® 755) [clone: LHM 2]
Anticorps
Type: Primary
Antigen: RNF5 (ring finger protein 5, E3 ubiquitin protein ligase)
Clonality: monoclonal
Clone: LHM 2
Conjugation: DyLight® 755
Epitope:
Host: Mouse
Isotype: IgG1 Kappa
Reactivity: Human
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR