Imprimer

Anti-Polyoma Virus, Medium T Antigen Rat Monoclonal Antibody (DyLight® 405) [clone: PyMT]

Fournisseur: Novus Biologicals
Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

NB100-2749V
NOVUNB100-2749VEA 667 EUR
NOVUNB100-2749V
Anti-Polyoma Virus, Medium T Antigen Rat Monoclonal Antibody (DyLight® 405) [clone: PyMT]
Anticorps
Type: Primary
Antigen: Polyoma Virus, Medium T Antigen
Clonality: monoclonal
Clone: PyMT
Conjugation: DyLight® 405
Epitope:
Host: Rat
Isotype: IgG2b
Reactivity: Virus
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR