Imprimer

Anti-CERK Rabbit Polyclonal Antibody

Fournisseur: Novus Biologicals
Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

NB100-2911 NB100-2911SS
NOVUNB100-2911EA 542 EUR
NOVUNB100-2911 NOVUNB100-2911SS
Anti-CERK Rabbit Polyclonal Antibody
Anticorps
Type: Primary
Antigen: CERK (ceramide kinase)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR