Imprimer

Anti-ATM Mouse Monoclonal Antibody (DyLight® 680) [clone: 10H11.E12]

Fournisseur: Novus Biologicals
Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

NB100-306FR
NOVUNB100-306FREA 638 EUR
NOVUNB100-306FR
Anti-ATM Mouse Monoclonal Antibody (DyLight® 680) [clone: 10H11.E12]
Anticorps
Type: Primary
Antigen: ATM (ataxia telangiectasia mutated)
Clonality: monoclonal
Clone: 10H11.E12
Conjugation: DyLight® 680
Epitope:
Host: Mouse
Isotype: IgG1 Kappa
Reactivity: Human, Mouse, Rat
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR