Imprimer

Anti-BUB1B Mouse Monoclonal Antibody (DyLight® 680) [clone: 8G1]

Fournisseur: Novus Biologicals
Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

NB100-353FR
NOVUNB100-353FREA 709 EUR
NOVUNB100-353FR
Anti-BUB1B Mouse Monoclonal Antibody (DyLight® 680) [clone: 8G1]
Anticorps
Type: Primary
Antigen: BUB1B (BUB1 mitotic checkpoint serine/threonine kinase B)
Clonality: monoclonal
Clone: 8G1
Conjugation: DyLight® 680
Epitope:
Host: Mouse
Isotype: IgG1 Kappa
Reactivity: Human, Mouse
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR