Imprimer

Anti-RPE65 Mouse Monoclonal Antibody (DyLight® 680) [clone: 401.8B11.3D9]

Fournisseur: Novus Biologicals
Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

NOVUNB100-355FREA 719 EUR
NOVUNB100-355FR
Anti-RPE65 Mouse Monoclonal Antibody (DyLight® 680) [clone: 401.8B11.3D9]
Anticorps
Type: Primary
Antigen: RPE65 (retinal pigment epithelium-specific protein 65kDa)
Clonality: monoclonal
Clone: 401.8B11.3D9
Conjugation: DyLight® 680
Epitope:
Host: Mouse
Isotype: IgG1 Kappa
Reactivity:
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR